siRNA Details
Gene Symbol: Olr1186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000862.1 | NP_001000862.1 | 1 | 454 |
Gene Symbol: LOC100911670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766038.1 | XP_008764260.1 | 454 |
Gene Symbol: LOC103693026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766037.1 | XP_008764259.1 | 1 | 127 |
