siRNA Details
Gene Symbol: Olr1142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000877.1 | NP_001000877.1 | 1 | 645 |
Gene Symbol: Olr1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001017.1 | NP_001001017.1 | 1 | 645 |
