siRNA Details
Gene Symbol: Olr1118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000883.1 | NP_001000883.1 | 1 | 400 |
Gene Symbol: LOC103693021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766027.1 | XP_008764249.1 | 2 | 163 |
