siRNA Details
Gene Symbol: Olr1366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000980.1 | NP_001000980.1 | 1 | 274 | ||
XM_008767521.1 | XP_008765743.1 | 2 | 424 | |||
XM_008767522.1 | XP_008765744.1 | 2 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
