siRNA Details
Gene Symbol: Olr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000997.1 | NP_001000997.1 | 1 | 331 | ||
XM_008762012.1 | XP_008760234.1 | 1 | 331 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000997.1 | NP_001000997.1 | 1 | 331 | ||
XM_008762012.1 | XP_008760234.1 | 1 | 331 |