siRNA Details
Gene Symbol: Olr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000997.1 | NP_001000997.1 | 1 | 62 | ||
XM_008762012.1 | XP_008760234.1 | 1 | 62 |

More Information
Other Species Targeted

Gene Ontology Categories:
