siRNA Details
Gene Symbol: Olr1681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001006.1 | NP_001001006.1 | 1 | 672 |
Gene Symbol: Olr1678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000893.1 | NP_001000893.1 | 1 | 672 |
Gene Symbol: Olr1683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000269.1 | NP_001000269.1 | 1 | 672 |

More Information
Other Species Targeted

Gene Ontology Categories:
