siRNA Details
Gene Symbol: Olr129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001027.1 | NP_001001027.1 | 1 | 412 |
Gene Symbol: Olr130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001272.1 | NP_001001272.1 | 1 | 412 |
