siRNA Details
Gene Symbol: Olr1395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001092.1 | NP_001001092.1 | 1 | 626 |
Gene Symbol: Olr1394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001091.1 | NP_001001091.1 | 1 | 626 |
Gene Symbol: Olr1396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001093.1 | NP_001001093.1 | 1 | 626 |

More Information
Other Species Targeted

Gene Ontology Categories:
