siRNA Details
Gene Symbol: Olr1397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001094.1 | NP_001001094.1 | 1 | 256 | ||
XM_008767693.1 | XP_008765915.1 | 4 | 1291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
