siRNA Details
Gene Symbol: Olr1462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001097.1 | NP_001001097.1 | 1 | 859 | ||
XM_006246511.2 | XP_006246573.1 | 2 | 963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
