siRNA Details
Gene Symbol: Olr1532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001103.1 | NP_001001103.1 | 1 | 640 |
Gene Symbol: Olr1529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001100.1 | NP_001001100.1 | 1 | 640 | ||
XM_006248195.2 | XP_006248257.1 | 2 | 739 |
Gene Symbol: Olr1530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001101.1 | NP_001001101.1 | 1 | 640 | ||
GenBank mRNA | AF091575.1 | - | 1 | 454 |
Gene Symbol: Olr1531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001102.1 | NP_001001102.1 | 1 | 640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
