siRNA Details
Gene Symbol: Olr1532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001103.1 | NP_001001103.1 | 1 | 620 |
Gene Symbol: Olr1530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001101.1 | NP_001001101.1 | 1 | 620 | ||
GenBank mRNA | AF091575.1 | - | 1 | 434 |
Gene Symbol: Olr1531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001102.1 | NP_001001102.1 | 1 | 620 |

More Information
Other Species Targeted

Gene Ontology Categories:
