siRNA Details
Gene Symbol: Olr1701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001114.1 | NP_001001114.1 | 1 | 784 | ||
XM_006255861.2 | XP_006255923.1 | 2 | 975 | |||
XM_006255863.2 | XP_006255925.1 | 2 | 944 | |||
XM_006255865.2 | XP_006255927.1 | 1 | 790 |
Gene Symbol: LOC100910739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751909.2 | XP_003751957.1 | 1 | 595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
