siRNA Details
Gene Symbol: Btnl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212488.1 | NP_997653.1 | 1711 | |||
XM_008772737.1 | XP_008770959.1 | 1711 |
Gene Symbol: LOC100911983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772783.1 | XP_008771005.1 | 10 | 1900 |
