siRNA Details
Gene Symbol: RT1-T18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002821.1 | NP_001002821.1 | 452 | |||
XM_006255985.2 | XP_006256047.1 | 3 | 1019 | |||
XM_006255986.2 | XP_006256048.1 | 3 | 1019 | |||
XM_008772742.1 | XP_008770964.1 | 3 | 1019 | |||
XM_008772743.1 | XP_008770965.1 | 4 | 484 | |||
XM_008772744.1 | XP_008770966.1 | 4 | 1622 | |||
GenBank mRNA | AY144125.1 | - | 373 | |||
BC104719.1 | - | 857 |
Gene Symbol: LOC102552616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256027.2 | XP_006256089.1 | 3 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
