siRNA Details
Gene Symbol: Tcf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213561.3 | NP_998726.2 | 604 | |||
GenBank mRNA | BC093374.1 | - | 531 | 7388599 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tcf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213561.3 | NP_998726.2 | 604 | |||
GenBank mRNA | BC093374.1 | - | 531 | 7388599 |