siRNA Details
Gene Symbol: G6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003976.1 | NP_001003976.1 | 2 | 349 | ||
XM_006256069.2 | XP_006256131.1 | 2 | 901 | |||
XM_008772746.1 | XP_008770968.1 | 2 | 901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
