siRNA Details
Gene Symbol: Ly6g5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001934.1 | NP_001001934.1 | 3 | 260 | ||
XM_008772747.1 | XP_008770969.1 | 4 | 947 | |||
GenBank mRNA | HE864428.1 | - | 2 | 171 | ||
HE864429.1 | - | 3 | 300 |
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HE864431.1 | - | 543 | |||
HE864432.1 | - | 1692 | ||||
HE864433.1 | - | 1917 | ||||
HE864434.1 | - | 2173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
