siRNA Details
Gene Symbol: Btla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213630.1 | NP_998795.1 | 2 | 123 | ||
XM_006248341.2 | XP_006248403.1 | 2 | 358 | |||
GenBank mRNA | AY590499.1 | - | 2 | 123 | ||
BC092588.1 | - | 2 | 327 | 7368263 | ||
DQ198369.1 | - | 2 | 123 |
