siRNA Details
Gene Symbol: Wfdc15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009179.1 | NP_001009179.1 | 2 | 167 | ||
XM_006235606.2 | XP_006235668.1 | 4 | 978 | |||
XM_006235607.2 | XP_006235669.1 | 3 | 1243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
