siRNA Details
Gene Symbol: Prss46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008865.1 | NP_001008865.1 | 3 | 392 | ||
XM_006243993.2 | XP_006244055.1 | 4 | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
