siRNA Details
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100781.1 | NP_001094251.1 | 2 | 403 | ||
XM_006237310.2 | XP_006237372.1 | 2 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
