siRNA Details
Gene Symbol: RT1-T24-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166403.1 | NP_001159875.1 | 2 | 149 | ||
XM_008772748.1 | XP_008770970.1 | 2 | 527 | |||
XM_008772749.1 | XP_008770971.1 | 2 | 527 | |||
XM_008772750.1 | XP_008770972.1 | 2 | 821 | |||
XM_008772751.1 | XP_008770973.1 | 3 | 612 | |||
XM_008772752.1 | XP_008770974.1 | 2 | 525 | |||
XM_008772753.1 | XP_008770975.1 | 2 | 525 | |||
GenBank mRNA | FQ230636.1 | - | 2 | 180 | ||
FQ234092.1 | - | 2 | 180 |
Gene Symbol: LOC100909797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772768.1 | XP_008770990.1 | 2 | 405 |
Gene Symbol: LOC360231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L40364.1 | - | 2 | 188 |
Gene Symbol: RT1-T24-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008826.3 | NP_001008826.2 | 2 | 232 | ||
GenBank mRNA | BC098866.1 | - | 2 | 187 | 7380738 | |
BC158853.1 | - | 188 | 7936214 | |||
FM045430.1 | - | 2 | 232 | |||
FQ213879.1 | - | 2 | 205 | |||
FQ225259.1 | - | 2 | 205 | |||
FQ225970.1 | - | 2 | 216 | |||
FQ233711.1 | - | 2 | 207 | |||
FQ234522.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
