siRNA Details
Gene Symbol: RT1-CE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008839.1 | NP_001008839.1 | 5 | 931 | ||
GenBank mRNA | BC090028.1 | - | 4 | 863 | 7365633 | |
U50449.1 | - | 5 | 922 |
Gene Symbol: LOC103690108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772769.1 | XP_008770991.1 | 4 | 1315 |
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ004889.1 | - | 4 | 859 | ||
AJ249698.1 | - | 6 | 859 | |||
AJ249704.1 | - | 5 | 859 | |||
L26224.1 | - | 5 | 940 | |||
L40363.1 | - | 6 | 919 | |||
M11071.1 | - | 2 | 149 | |||
M24023.1 | - | 4 | 709 | |||
M24024.1 | - | 3 | 408 | |||
U38970.1 | - | 5 | 922 | |||
X90370.1 | - | 4 | 859 | |||
X99767.1 | - | 4 | 859 | |||
Y08530.1 | - | 4 | 859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
