siRNA Details
Gene Symbol: Dnmt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003957.1 | NP_001003957.1 | 6 | 773 | ||
NM_001003958.1 | NP_001003958.1 | 10 | 1455 | |||
XM_008764530.1 | XP_008762752.1 | 9 | 1314 | |||
XM_008764531.1 | XP_008762753.1 | 6 | 908 |

More Information
Other Species Targeted

Gene Ontology Categories:
