siRNA Details
Gene Symbol: Reg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004096.1 | NP_001004096.1 | 1, 2 | 72 | ||
XM_006233009.2 | XP_006233071.1 | 2, 3 | 275 | |||
XM_008761321.1 | XP_008759543.1 | 2, 3 | 238 | |||
GenBank mRNA | AB164049.1 | - | 1, 2 | 72 |
