siRNA Details
Gene Symbol: Ly96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024279.1 | NP_001019450.1 | 212 | |||
GenBank mRNA | AY197554.1 | - | 157 | |||
AY963291.1 | - | 212 |

More Information
Other Species Targeted

Gene Ontology Categories:
