siRNA Details
Gene Symbol: Emr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007558.1 | NP_001007559.1 | 1985 | |||
XM_008766767.1 | XP_008764989.1 | 1973 | ||||
GenBank mRNA | AY686633.1 | - | 1985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
