siRNA Details
Gene Symbol: Emr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007558.1 | NP_001007559.1 | 111 | |||
XM_008766767.1 | XP_008764989.1 | 105 | ||||
GenBank mRNA | AY686633.1 | - | 111 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
