siRNA Details
Gene Symbol: Clec4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005899.2 | NP_001005899.2 | 4 | 373 | ||
GenBank mRNA | AY363176.1 | - | 4 | 419 | ||
AY494060.1 | - | 4 | 372 | |||
AY581051.1 | - | 4 | 373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
