siRNA Details
Gene Symbol: Sspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007016.1 | NP_001007017.1 | 15, 16 | 2321 | ||
XM_008762902.1 | XP_008761124.1 | 16, 17 | 3822 | |||
GenBank mRNA | AJ629845.1 | - | 15, 16 | 2321 |
