siRNA Details
Gene Symbol: Sspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007016.1 | NP_001007017.1 | 94 | 14402 | ||
XM_008762902.1 | XP_008761124.1 | 95 | 15873 | |||
GenBank mRNA | AJ629845.1 | - | 94 | 14402 |
