siRNA Details
Gene Symbol: Pmaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008385.1 | NP_001008386.1 | 2 | 94 | ||
GenBank mRNA | AY788892.1 | - | 2 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
