siRNA Details
Gene Symbol: Klra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009486.1 | NP_001009486.1 | 6 | 647 | ||
XM_008763404.1 | XP_008761626.1 | 5 | 624 | |||
GenBank mRNA | AY649835.1 | - | 6 | 647 | ||
BC085917.1 | - | 610 | 7108565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
