siRNA Details
Gene Symbol: Ly49s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009488.1 | NP_001009488.1 | 6 | 654 | ||
GenBank mRNA | AY649837.1 | - | 6 | 654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
