siRNA Details
Gene Symbol: Ly49i4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009495.1 | NP_001009495.1 | 1 | 173 | ||
XM_008763405.1 | XP_008761627.1 | 2 | 288 | |||
GenBank mRNA | AY649834.1 | - | 1 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
