siRNA Details
Gene Symbol: Ly49i9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009496.1 | NP_001009496.1 | 4 | 447 | ||
GenBank mRNA | AY649839.1 | - | 4 | 447 |
Gene Symbol: LOC497796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111310.1 | NP_001104780.1 | 440 | |||
XM_006237126.2 | XP_006237188.1 | 4 | 490 | |||
GenBank mRNA | BC093399.1 | - | 440 | 7389533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
