siRNA Details
Gene Symbol: Ly49si1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009497.1 | NP_001009497.1 | 6 | 711 | ||
GenBank mRNA | AY659935.1 | - | 6 | 711 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
