siRNA Details
Gene Symbol: Ly49i5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009501.2 | NP_001009501.1 | 734 | |||
GenBank mRNA | AY649836.1 | - | 734 | |||
AY653729.1 | - | 579 | ||||
AY846264.1 | - | 584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
