siRNA Details
Gene Symbol: Pfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009503.1 | NP_001009503.1 | 2 | 1019 | ||
GenBank mRNA | AY682392.1 | - | 2 | 1019 | ||
BC107465.1 | - | 2 | 1151 | 7453892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
