siRNA Details
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009504.1 | NP_001009504.1 | 1 | 539 | ||
XM_006246020.2 | XP_006246082.1 | 2 | 857 | |||
GenBank mRNA | AY823669.1 | - | 1 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
