siRNA Details
Gene Symbol: LOC494224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009505.2 | NP_001009505.1 | 4 | 228 | ||
XM_006250794.2 | XP_006250856.1 | 3 | 369 | |||
GenBank mRNA | AB196306.1 | - | 4 | 228 |
