siRNA Details
Gene Symbol: Vom1r111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009522.1 | NP_001009522.1 | 1 | 372 | ||
GenBank mRNA | AY510366.1 | - | 1 | 372 |
Gene Symbol: Vom1r17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167572.1 | NP_001161044.1 | 372 |
