siRNA Details
Gene Symbol: Vom1r59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008944.1 | NP_001008944.1 | 1 | 773 | ||
GenBank mRNA | AY510282.1 | - | 1 | 773 |
Gene Symbol: LOC103691064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759093.1 | XP_008757315.1 | 455 |
Gene Symbol: Vom1r60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167552.1 | NP_001161024.1 | 2 | 773 |
