siRNA Details
Gene Symbol: Vom1r81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008970.1 | NP_001008970.1 | 1 | 212 | ||
GenBank mRNA | AY510288.1 | - | 1 | 212 |
Gene Symbol: LOC100909473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236570.1 | XP_006236632.1 | 1 | 212 |
Gene Symbol: Vom1r75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008972.1 | NP_001008972.1 | 1 | 212 | ||
GenBank mRNA | AY510283.1 | - | 1 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
