siRNA Details
Gene Symbol: Scgb2b24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_577783.2 | XP_577783.2 | 3 | 271 | ||
GenBank mRNA | AY871782.1 | - | 2 | 202 | ||
GU269242.1 | - | 3 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
