siRNA Details
Gene Symbol: LRRFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137552.1 | NP_001131024.1 | 11 | 2340 | ||
NM_001137553.1 | NP_001131025.1 | 8 | 2172 | |||
NM_004735.3 | NP_004726.2 | 10 | 2268 | |||
GenBank mRNA | AJ223075.1 | - | 10 | 2154 | ||
BC108913.1 | - | 8 | 1997 | 40027210 | ||
BC108914.1 | - | 11 | 2165 | 40027218 | ||
KP455483.1 | - | 8 | 1880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
