siRNA Details
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012514.1 | NP_036646.1 | 10 | 2593 | ||
XM_008768092.1 | XP_008766314.1 | 10 | 2809 | |||
GenBank mRNA | AF036760.1 | - | 10 | 2593 |
