siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040271.1 | NP_001035361.1 | 4 | 360 | ||
NM_053592.2 | NP_446044.1 | 5 | 480 | |||
XM_008762188.1 | XP_008760410.1 | 5 | 518 | |||
GenBank mRNA | CB325379.1 | - | 4 | 374 | ||
FQ220451.1 | - | 515 | ||||
U64030.2 | - | 5 | 480 |
